ENA432539543 Enamine

2-{[(5-amino-2-chlorophenyl)methyl](methyl)amino}ethan-1-ol

  • MDL number MFCD12813435
  • Cas Number : 1179853-30-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196100

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196100
Availability details for
Quantity
  • Availability
  • Add to Cart