ENA432539544 Enamine

1-{bicyclo[4.2.0]octa-1(6),2,4-trien-7-yl}-2-bromoethan-1-one

  • MDL number MFCD23845004
  • Cas Number : 1499640-27-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-195073

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195073
Availability details for
Quantity
  • Availability
  • Add to Cart