ENA432539550 Enamine

ethyl 3-amino-2-phenylpyridine-4-carboxylate

  • MDL number MFCD25250001
  • Cas Number : 1555792-30-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194898

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194898
Availability details for
Quantity
  • Availability
  • Add to Cart