ENA432539554 Enamine

N4-cyclopentyl-N4-ethylpyrimidine-2,4-diamine

  • MDL number MFCD23985604
  • Cas Number : 1512107-90-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195080

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195080
Availability details for
Quantity
  • Availability
  • Add to Cart