ENA432539555 Enamine

3-amino-2-phenylpyridine-4-carboxylic acid

  • MDL number MFCD24264511
  • Cas Number : 1547501-48-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194899

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194899
Availability details for
Quantity
  • Availability
  • Add to Cart