ENA432539557 Enamine

(1R)-1-(4-ethylphenyl)ethan-1-ol

  • MDL number MFCD09863724
  • Cas Number : 54225-75-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196139
Availability details for
Quantity
  • Availability
  • Add to Cart