ENA432539558 Enamine

N,3-dimethylpyrrolidine-3-carboxamide

  • MDL number MFCD18273671
  • Cas Number : 1342746-44-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196140

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196140
Availability details for
Quantity
  • Availability
  • Add to Cart