ENA432539559 Enamine

methyl 5,6-diaminopyridine-2-carboxylate hydrochloride

  • MDL number MFCD18070974
  • Cas Number : 1803600-70-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196141
Availability details for
Quantity
  • Availability
  • Add to Cart