ENA432539561 Enamine

(4aR,7aS)-6-benzyl-octahydro-1H-pyrrolo[3,4-b]pyridine-5,7-dione

  • MDL number MFCD28656621
  • Cas Number : 151213-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196148
Availability details for
Quantity
  • Availability
  • Add to Cart