ENA432539564 Enamine

2-(3-aminobenzenesulfinyl)-N-ethylacetamide

  • MDL number MFCD24062575
  • Cas Number : 1510580-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196158

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196158
Availability details for
Quantity
  • Availability
  • Add to Cart