ENA432539565 Enamine

4-(1H-1,2,3,4-tetrazol-5-yl)piperidin-4-ol hydrochloride

  • MDL number MFCD28145318
  • Cas Number : 1803589-30-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196160

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196160
Availability details for
Quantity
  • Availability
  • Add to Cart