ENA432539566 Enamine

ethyl 2-amino-3-(3-chlorophenyl)propanoate hydrochloride

  • MDL number MFCD28145319
  • Cas Number : 457654-55-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-141
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196161

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196161
Availability details for
Quantity
  • Availability
  • Add to Cart