ENA432539568 Enamine

N-cyclopropylpyridin-4-amine

  • MDL number MFCD11118361
  • Cas Number : 1036598-73-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196167

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196167
Availability details for
Quantity
  • Availability
  • Add to Cart