ENA432539571 Enamine

2-(bromomethyl)-4-chloro-1-(cyclopropylmethoxy)benzene

  • MDL number MFCD12138930
  • Cas Number : 1103261-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195084
Availability details for
Quantity
  • Availability
  • Add to Cart