ENA432539580 Enamine

4-chloro-7-fluoro-6-methoxyquinoline

  • MDL number MFCD15527286
  • Cas Number : 25759-94-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 153-154
Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195097

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195097
Availability details for
Quantity
  • Availability
  • Add to Cart