ENA432539584 Enamine

ethyl 2-hydroxy-2-(4-methanesulfonylphenyl)propanoate

  • MDL number MFCD17317128
  • Cas Number : 1275811-23-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196225

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196225
Availability details for
Quantity
  • Availability
  • Add to Cart