ENA432539585 Enamine

1-(3-amino-4-methylphenyl)-3,3-dimethylurea

  • MDL number MFCD21809479
  • Cas Number : 1509056-86-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196227

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196227
Availability details for
Quantity
  • Availability
  • Add to Cart