ENA432539586 Enamine

5-(chloromethyl)-2-methoxy-4-methylpyrimidine

  • MDL number MFCD26013984
  • Cas Number : 1555080-80-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196228

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196228
Availability details for
Quantity
  • Availability
  • Add to Cart