ENA432539589 Enamine

3-[(2,5-difluorophenyl)sulfanyl]prop-2-enoic acid

  • MDL number MFCD23791353
  • Cas Number : 1564084-66-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-196232
Availability details for
Quantity
  • Availability
  • Add to Cart