ENA432539590 Enamine

1-(5-bromopyridin-2-yl)piperidine-3-carboxamide

  • MDL number MFCD14650338
  • Cas Number : 1250050-53-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196234
Availability details for
Quantity
  • Availability
  • Add to Cart