ENA432539591 Enamine

5-bromo-N-[3-(dimethylamino)-2,2-dimethylpropyl]pyridin-2-amine

  • MDL number MFCD11125301
  • Cas Number : 1040328-99-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196235
Availability details for
Quantity
  • Availability
  • Add to Cart