ENA432539594 Enamine

[(4-chlorophenyl)methyl][(1-methyl-1H-pyrazol-4-yl)methyl]amine

  • MDL number MFCD11167547
  • Cas Number : 1152953-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196238

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196238
Availability details for
Quantity
  • Availability
  • Add to Cart