ENA432539597 Enamine

4-chloro-3-[(morpholin-4-yl)methyl]aniline

  • MDL number MFCD10021403
  • Cas Number : 769961-16-2
  • Molecular Weight : 226.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-chloro-3-(4-morpholinylmethyl)aniline
Inchi Code 1S/C11H15ClN2O/c12-11-2-1-10(13)7-9(11)8-14-3-5-15-6-4-14/h1-2,7H,3-6,8,13H2
Inchi Key KHBXNTAWKWNUIS-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196245

Safety Information

signalword Danger
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P310,P312,P330,P332+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-196245
pictograms GHS05,GHS07
hstatements H302,H312,H315,H318,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart