ENA432539598 Enamine

methyl 2-formylthiophene-3-carboxylate

  • MDL number MFCD01859943
  • Cas Number : 24647-82-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196247

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196247
Availability details for
Quantity
  • Availability
  • Add to Cart