ENA432539605 Enamine

6-(2,6-difluorobenzoyl)-2,3-dihydro-1,4-benzodioxine

  • MDL number MFCD13660710
  • Cas Number : 1097078-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196268

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196268
Availability details for
Quantity
  • Availability
  • Add to Cart