ENA432539606 Enamine

(4-chlorophenyl)(cyclopentyl)methanone

  • MDL number MFCD03841308
  • Cas Number : 2204-98-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196270

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196270
Availability details for
Quantity
  • Availability
  • Add to Cart