ENA432539608 Enamine

hexane-3-sulfonyl chloride

  • MDL number MFCD19200632
  • Cas Number : 1081524-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195106

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195106
Availability details for
Quantity
  • Availability
  • Add to Cart