ENA432539610 Enamine

1-(1,3-thiazol-2-yl)ethan-1-amine hydrochloride

  • MDL number MFCD11846195
  • Cas Number : 947662-64-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196290
Availability details for
Quantity
  • Availability
  • Add to Cart