ENA432539611 Enamine

1-(pyridin-4-yl)-1H,4H,5H-pyrazolo[3,4-d]pyrimidin-4-one hydrochloride

  • MDL number MFCD28145336
  • Cas Number : 1803583-22-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196292
Availability details for
Quantity
  • Availability
  • Add to Cart