ENA432539613 Enamine

2-chloro-N-methyl-5-propanoylbenzene-1-sulfonamide

  • MDL number MFCD11925251
  • Cas Number : 1175862-48-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195109

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195109
Availability details for
Quantity
  • Availability
  • Add to Cart