ENA432539614 Enamine

2-methyl-4,5,6,7-tetrahydro-2H-indazol-4-one

  • MDL number MFCD11847436
  • Cas Number : 1027617-67-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194900
Availability details for
Quantity
  • Availability
  • Add to Cart