ENA432539617 Enamine

ethyl 2-amino-5-bromo-3-ethylbenzoate

  • MDL number MFCD00229180
  • Cas Number : 34921-62-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 37-38
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195111

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195111
Availability details for
Quantity
  • Availability
  • Add to Cart