ENA432539618 Enamine

5-(cyclopentanesulfonyl)-2-fluorobenzoic acid

  • MDL number MFCD11539751
  • Cas Number : 1097007-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-195112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195112
Availability details for
Quantity
  • Availability
  • Add to Cart