ENA432539625 Enamine

5-bromo-N-[2-(diethylamino)ethyl]pyridin-2-amine

  • MDL number MFCD11124557
  • Cas Number : 91250-98-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196366

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196366
Availability details for
Quantity
  • Availability
  • Add to Cart