ENA432539627 Enamine

1-(2,5-difluoro-4-methoxyphenyl)-5-propyl-1H-pyrazol-4-amine hydrochloride

  • MDL number MFCD28139596
  • Cas Number : 1803588-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195136

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195136
Availability details for
Quantity
  • Availability
  • Add to Cart