ENA432539630 Enamine

5-[(pyrrolidin-1-yl)methyl]pyridin-2-amine

  • MDL number MFCD16689272
  • Cas Number : 913573-02-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196371

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196371
Availability details for
Quantity
  • Availability
  • Add to Cart