ENA432539632 Enamine

5-methyl-1,2,3,6-tetrahydropyridine hydrochloride

  • MDL number MFCD27988116
  • Cas Number : 1523617-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196374

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196374
Availability details for
Quantity
  • Availability
  • Add to Cart