ENA432539638 Enamine

2-[(4-bromo-2-methylphenyl)amino]acetic acid

  • MDL number MFCD06849859
  • Cas Number : 66947-37-3
  • Molecular Weight : 244.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromo-2-methylanilino)acetic acid
Inchi Code 1S/C9H10BrNO2/c1-6-4-7(10)2-3-8(6)11-5-9(12)13/h2-4,11H,5H2,1H3,(H,12,13)
Inchi Key VLQCUVXTNBDRRF-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-196381

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196381
Availability details for
Quantity
  • Availability
  • Add to Cart