ENA432539639 Enamine

(3-methyl-1,2,4-oxadiazol-5-yl)(oxan-4-yl)methanamine hydrochloride

  • MDL number MFCD28139599
  • Cas Number : 1798767-40-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195139
Availability details for
Quantity
  • Availability
  • Add to Cart