ENA432539640 Enamine

2-hydroxy-5-iodo-4-methylpyridine-3-carbonitrile

  • MDL number MFCD09834636
  • Cas Number : 1378640-39-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196387

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196387
Availability details for
Quantity
  • Availability
  • Add to Cart