ENA432539641 Enamine

6,8-dichloroimidazo[1,2-a]pyrazine

  • MDL number MFCD18072707
  • Cas Number : 1245645-38-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 115-116
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196389

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196389
Availability details for
Quantity
  • Availability
  • Add to Cart