ENA432539647 Enamine

5-fluoro-2-(1-methyl-1H-pyrazol-5-yl)phenol

  • MDL number MFCD25994628
  • Cas Number : 1547300-43-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-195143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195143
Availability details for
Quantity
  • Availability
  • Add to Cart