ENA432539650 Enamine

1-[5-chloro-2-(morpholin-4-yl)phenyl]methanamine

  • MDL number MFCD16769528
  • Cas Number : 1250339-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196473
Availability details for
Quantity
  • Availability
  • Add to Cart