ENA432539654 Enamine

7-bromo-3-chloro-2-methyl-4H-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD02954461
  • Cas Number : 354792-26-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-194905

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-194905
Availability details for
Quantity
  • Availability
  • Add to Cart