ENA432539655 Enamine

6-(2-amino-1,3-thiazol-4-yl)-7-fluoro-1,2,3,4-tetrahydroquinolin-2-one

  • MDL number MFCD11981529
  • Cas Number : 1154114-88-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196496

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196496
Availability details for
Quantity
  • Availability
  • Add to Cart