ENA432539662 Enamine

3-{[(3-aminophenyl)carbamoyl]amino}propanamide

  • MDL number MFCD12136700
  • Cas Number : 1157041-14-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196551

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196551
Availability details for
Quantity
  • Availability
  • Add to Cart