ENA432539665 Enamine

[(4-bromophenyl)methyl](octan-2-yl)amine

  • MDL number MFCD12646072
  • Cas Number : 1152862-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196558

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196558
Availability details for
Quantity
  • Availability
  • Add to Cart