ENA432539686 Enamine

spiro[imidazolidine-4,8'-tricyclo[5.2.1.0,2,6]decane]-2,5-dione

  • MDL number MFCD17237707
  • Cas Number : 59455-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-196583

Safety Information

signalword Warning
pcodes P261,P264,P270,P272,P280,P301+P312,P302+P352,P330,P333+P313,P363,P501
hcodes Xn
MSDS https://www.enaminestore.com/catalog/EN300-196583
pictograms GHS07
hstatements H302,H317
Availability details for
Quantity
  • Availability
  • Add to Cart