ENA432539698 Enamine

(3-bromopropyl)(methyl)amine hydrobromide

  • MDL number MFCD27954546
  • Cas Number : 60035-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 64-66
Purity 95
Salt Data HBr
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-196617

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196617
Availability details for
Quantity
  • Availability
  • Add to Cart