ENA432539700 Enamine

2-(bromomethyl)pyrrolidine hydrobromide

  • MDL number MFCD20527247
  • Cas Number : 3433-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HBr
Storage temp. -20
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-196619

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196619
Availability details for
Quantity
  • Availability
  • Add to Cart